Methyl sulfoxide, 99.7+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Methyl sulfoxide, 99.7+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Methyl sulfoxide, 99.7+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Methyl sulfoxide, 99.7+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Methyl sulfoxide, 99.7+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfoxide, 99.7+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACO326881000100 mLAcroSeal™ Glass Bottle
Catalog number ACO326881000
Price (SGD)
125.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (SGD)
125.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethyl Sulfoxide
Melting Point18.4°C
CAS67-68-5
ColorColorless
Density1.1000g/mL