1x1 image pixel for data collection
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACO326871000
Price (SGD)
110.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (SGD)
110.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N-Dimethylformamide
Melting Point-61.0°C
CAS68-12-2
CAS Min %99.75
ColorColorless
View more
RUO – Research Use Only
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link