Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene
Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene
Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene
Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene
Thermo Scientific Chemicals
Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO32676010010 LMetal Drum
Catalog number ACO326760100
Price (SGD)
515.00
EA
Add to cart
Quantity:
10 L
Packaging:
Metal Drum
Request bulk or custom format
Price (SGD)
515.00
EA
Add to cart
Specifications
Chemical Name or MaterialDichloromethane
Melting Point-97.0°C
Absorbance0.5 max. at 235nm, 0.108 max. at 240nm, 0.009 max. at 260nm
CAS75-09-2
Health Hazard 1Warning
View more
For residue analysis
RUO – Research Use Only