Rhodium(III) acetylacetonate, 97%
Rhodium(III) acetylacetonate, 97%
Rhodium(III) acetylacetonate, 97%
Rhodium(III) acetylacetonate, 97%
Thermo Scientific Chemicals
Rhodium(III) acetylacetonate, 97%
CAS: 14284-92-5 | C15H21O6Rh | 400.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO325452500250 mgGlass bottle
Catalog number ACO325452500
Price (SGD)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialRhodium(III) acetylacetonate
CAS14284-92-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.