Copper(II) chloride dihydrate, 99+%, ACS reagent
Copper(II) chloride dihydrate, 99+%, ACS reagent
Copper(II) chloride dihydrate, 99+%, ACS reagent
Copper(II) chloride dihydrate, 99+%, ACS reagent
Thermo Scientific Chemicals
Copper(II) chloride dihydrate, 99+%, ACS reagent
CAS: 10125-13-0 | Cl2CuH4O2 | 170.48 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO3152800252.5 kgPlastic Bottle
Catalog number ACO315280025
Price (SGD)
1,100.00
EA
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (SGD)
1,100.00
EA
Add to cart
Specifications
Chemical Name or MaterialCopper(II) Chloride Dihydrate
Melting Point100.0°C
Name NoteFor analysis ACS
CAS10125-13-0