Tetracontane, 98%
Tetracontane, 98%
Tetracontane, 98%
Thermo Scientific Chemicals
Tetracontane, 98%
CAS: 4181-95-7 | C40H82 | 563.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO3091600252.5 gGlass bottle
Catalog number ACO309160025
Price (SGD)
487.00
EA
Add to cart
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
487.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetracontane
CAS4181-95-7
Melting Point80.0°C to 83.0°C
ColorWhite
EINECS Number224-055-7