1-Methoxy-2-propanol, 98.5%, extra pure
1-Methoxy-2-propanol, 98.5%, extra pure
1-Methoxy-2-propanol, 98.5%, extra pure
1-Methoxy-2-propanol, 98.5%, extra pure
Thermo Scientific Chemicals
1-Methoxy-2-propanol, 98.5%, extra pure
CAS: 107-98-2 | C4H10O2 | 90.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO2449900101 LGlass Bottle
Catalog number ACO244990010
Price (SGD)
112.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
112.00
EA
Add to cart
Specifications
Chemical Name or Material1-Methoxy-2-propanol
CAS107-98-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest