Polyvinylpyrrolidone, average M.W. 8.000, K16-18
Polyvinylpyrrolidone, average M.W. 8.000, K16-18
Polyvinylpyrrolidone, average M.W. 8.000, K16-18
Polyvinylpyrrolidone, average M.W. 8.000, K16-18
Thermo Scientific Chemicals
Polyvinylpyrrolidone, average M.W. 8.000, K16-18
CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO227465000500 gPlastic bottle
Catalog number ACO227465000
Price (SGD)
237.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
237.00
EA
Add to cart
Specifications
Chemical Name or MaterialPolyvinylpyrrolidone
ColorWhite or White-Yellow
Name NoteK16-18
pH3.0 to 5.0 (5% aq. soln.)
CAS9003-39-8