Triethylamine, 99.7%, extra pure
Triethylamine, 99.7%, extra pure
Triethylamine, 99.7%, extra pure
Triethylamine, 99.7%, extra pure
Thermo Scientific Chemicals
Triethylamine, 99.7%, extra pure
CAS: 121-44-8 | C6H15N | 101.19 g/mol
Have Questions?
Catalog NumberQuantity
ACO21951050050 g
Catalog number ACO219510500
Price (SGD)
51.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (SGD)
51.00
EA
Add to cart
Specifications
Chemical Name or MaterialTriethylamine
Name Note99.7%
CAS121-44-8
Health Hazard 1Danger
Health Hazard 2GHS P Statement Highly flammable liquid and vapour. Toxic if inhaled. Toxic in contact with skin. Harmful if swallowed. Causes severe skin burns and eye damage. May cause respiratory irritation.