Drierite™, without indicator, 8 mesh
Drierite™, without indicator, 8 mesh
Drierite™, without indicator, 8 mesh
Drierite™, without indicator, 8 mesh
Thermo Scientific Chemicals
Drierite™, without indicator, 8 mesh
CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO21905010010 kgPlastic Drum
Catalog number ACO219050100
Price (SGD)
776.00
EA
Add to cart
Quantity:
10 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (SGD)
776.00
EA
Add to cart
Specifications
Chemical Name or MaterialDrierite™
Melting Point1450.0°C
Physical FormGranular
CAS7778-18-9
Health Hazard 1
View more
Adsorbent, Indicator
RUO – Research Use Only