Linoleic acid, 99%
Linoleic acid, 99%
Linoleic acid, 99%
Thermo Scientific Chemicals
Linoleic acid, 99%
CAS: 60-33-3 | C18H32O2 | 280.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number ACO215041000
Price (SGD)
1,561.60
Special offer
Ends: 31-Mar-2026
1,561.60
Each
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
1,561.60
Special offer
Ends: 31-Mar-2026
1,561.60
Each
Add to cart
Specifications
Chemical Name or MaterialLinoleic acid
CAS60-33-3
Health Hazard 1Warning
Melting Point-5.0°C
ColorColorless to Yellow