Ammonium iron(III) citrate, 16.5 to 18.5% Fe
Ammonium iron(III) citrate, 16.5 to 18.5% Fe
Ammonium iron(III) citrate, 16.5 to 18.5% Fe
Ammonium iron(III) citrate, 16.5 to 18.5% Fe
Thermo Scientific Chemicals
Ammonium iron(III) citrate, 16.5 to 18.5% Fe
CAS: 1185-57-5 | C6H11FeNO7 | 265 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO2118400252.5 kgPlastic bottle
Catalog number ACO211840025
Price (SGD)
258.00
EA
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
258.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium iron(III) citrate
Name Note16.5 to 18.5% Fe
CAS1185-57-5
ColorBrown to Brown-Yellow
Health Hazard 1HYGROSCOPIC