Calcium chloride dihydrate, 99+%, for analysis
Calcium chloride dihydrate, 99+%, for analysis
Calcium chloride dihydrate, 99+%, for analysis
Calcium chloride dihydrate, 99+%, for analysis
Thermo Scientific Chemicals
Calcium chloride dihydrate, 99+%, for analysis
CAS: 10035-04-8 | CaCl2·2H2O | 147.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO2077800101 kgPlastic Bottle
Catalog number ACO207780010
Price (SGD)
153.60
Special offer
Ends: 31-Mar-2026
153.60
Each
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (SGD)
153.60
Special offer
Ends: 31-Mar-2026
153.60
Each
Add to cart
Specifications
Chemical Name or MaterialCalcium chloride dihydrate
Melting Point175.0°C
CAS10035-04-8
ColorWhite
Health Hazard 1GHS Signal Word: Warning
View more
Adsorbent
RUO – Research Use Only