Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water
Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACO205840010
Price (SGD)
63.20
Special offer
Ends: 31-Mar-2026
63.20
Each
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
63.20
Special offer
Ends: 31-Mar-2026
63.20
Each
Add to cart
Specifications
Chemical Name or MaterialAmmonium hydroxide
Boiling Point38.0°C
CAS1336-21-6
Health Hazard 1Danger
Melting Point-57.0°C