Sodium phosphate, dibasic, 99+%, for analysis, anhydrous
Sodium phosphate, dibasic, 99+%, for analysis, anhydrous
Sodium phosphate, dibasic, 99+%, for analysis, anhydrous
Sodium phosphate, dibasic, 99+%, for analysis, anhydrous
Thermo Scientific Chemicals
Sodium phosphate, dibasic, 99+%, for analysis, anhydrous
CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO204855000500 gPlastic bottle
Catalog number ACO204855000
Price (SGD)
122.40
Special offer
Ends: 31-Mar-2026
122.40
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
122.40
Special offer
Ends: 31-Mar-2026
122.40
Each
Add to cart
Specifications
Chemical Name or MaterialSodium phosphate, dibasic
Name Notep.a.
CAS7558-79-4
ColorWhite
Health Hazard 1HYGROSCOPIC
View more
Biological buffer for analysis
RUO – Research Use Only