Hexylamine, 99%
Hexylamine, 99%
Hexylamine, 99%
Thermo Scientific Chemicals
Hexylamine, 99%
CAS: 111-26-2 | C6H15N | 101.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass bottle
Catalog number ACO204721000
Price (SGD)
58.40
Special offer
Ends: 31-Mar-2026
58.40
Each
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
58.40
Special offer
Ends: 31-Mar-2026
58.40
Each
Add to cart
Specifications
Chemical Name or MaterialHexylamine
Name Note99%
CAS111-26-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if swallowed.
Harmful to aquatic life with long lasting effects.