Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized
CAS: 7722-84-1 | H2O2 | 34 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ACO202460010
1 L
Plastic bottle
Catalog number ACO202460010
Price (SGD)
124.80
Special offer
Ends: 31-Mar-2026
124.80
Each
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
124.80
Special offer
Ends: 31-Mar-2026
124.80
Each
Add to cart
Specifications
Boiling Point108.0°C
Chemical Name or MaterialHydrogen peroxide
Melting Point-33.0°C
Name NoteFor analysis, Stabilized
CAS7722-84-1