Lithium bromide, 99+%, for analysis, anhydrous
Lithium bromide, 99+%, for analysis, anhydrous
Lithium bromide, 99+%, for analysis, anhydrous
Lithium bromide, 99+%, for analysis, anhydrous
Thermo Scientific Chemicals
Lithium bromide, 99+%, for analysis, anhydrous
CAS: 7550-35-8 | BrLi | 86.84 g/mol
Have Questions?
Catalog NumberQuantity
ACO199875000500 g
Catalog number ACO199875000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point1265.0°C
Chemical Name or MaterialLithium bromide
Melting Point550.0°C
Name Note99+%
CAS7550-35-8