Poly(ethylene glycol), average M.W. 400
Poly(ethylene glycol), average M.W. 400
Poly(ethylene glycol), average M.W. 400
Poly(ethylene glycol), average M.W. 400
Thermo Scientific Chemicals
Poly(ethylene glycol), average M.W. 400
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1922300101 LGlass Bottle
Catalog number ACO192230010
Price (SGD)
149.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
149.00
EA
Add to cart
Specifications
Chemical Name or MaterialPoly(ethylene glycol)
ColorColorless
CAS25322-68-3
Physical FormLiquid
Boiling Point>200.0°C