Oil Red O, Biological Stain
Oil Red O, Biological Stain
Oil Red O, Biological Stain
Oil Red O, Biological Stain
Thermo Scientific Chemicals
Oil Red O, Biological Stain
CAS: 1320-06-5 | C26H24N4O | 408.51 g/mol
Have Questions?
Catalog NumberQuantity
ACO18940025025 g
Catalog number ACO189400250
Price (SGD)
50.40
Special offer
Ends: 31-Mar-2026
63.00
Save 12.60 (20%)
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
50.40
Special offer
Ends: 31-Mar-2026
63.00
Save 12.60 (20%)
EA
Add to cart
Specifications
Chemical Name or MaterialOil Red O
CAS1320-06-5
Molecular FormulaC26H24N4O
MDL NumberMFCD00003898
SynonymC.I. 26125, Solvent Red 27