1H-Benzotriazole, 99%
1H-Benzotriazole, 99%
1H-Benzotriazole, 99%
1H-Benzotriazole, 99%
Thermo Scientific Chemicals
1H-Benzotriazole, 99%
CAS: 95-14-7 | C6H5N3 | 119.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO183885000500 gPlastic bottle
Catalog number ACO183885000
Price (SGD)
240.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
240.00
EA
Add to cart
Specifications
Chemical Name or Material1H-Benzotriazole
Name Note98%
CAS95-14-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
View more
Indicator
RUO – Research Use Only