Nalidixic acid, 99.5%
Nalidixic acid, 99.5%
Nalidixic acid, 99.5%
Nalidixic acid, 99.5%
Thermo Scientific Chemicals
Nalidixic acid, 99.5%
Nalixidic acid, or nalixidin, CAS # 389-08-2, is a quinolone antibiotic used to inhibit DNA gyrase and prevent cell division. | CAS: 389-08-2 | C12H12N2O3 | 232.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1699000505 gGlass bottle
Catalog number ACO169900050
Price (SGD)
59.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
59.00
EA
Add to cart
Specifications
Chemical Name or MaterialNalidixic acid
Name Note99.5%
CAS389-08-2
CAS Min %99.4
Health Hazard 1GHS Signal Word: Danger
View more
General Description
• Nalixidic acid is a synthetic quinolone that is a monocarboxylic acid composed of 1,8-naphthyridine substituted with carboxylic acid, ethyl, and methyl groups
• Nalixidic acid is a DNA gyrase inhibitor that prevents DNA synthesis and subsequent cell divisions
Applications
• Nalixidic acid is primarily effective against gram-negative bacteria
• It may be useful for bacterial resistance studies and laboratory research on drug allergies and cross-reactivity
• Nalixidic acid is suitable for high-performance liquid chromatography (HPLC) and related biochemistry laboratory applications
RUO – Research Use Only
General References:
  1. Kumar, Y.; Sharma, A.; Mani, K.R. Characterization of antimicrobial resistance markers & their stability in Salmonella enterica serovar Typhi. Indian J Med Res. 2017, 146, S9-S14.
  2. Morales-Irigoyen, E.E.; de Las Mercedes Gómez-Y-Gómez, Y.; Flores-Moreno, J.L.; Franco-Hernández, M.O. A bionanohybrid ZnAl-NADS ecological pesticide as a treatment for soft rot disease in potato (Solanum tuberosum L.). Environ Sci Pollut Res Int. 2018, 25(22), 21430-21439.