1,3,5-Benzenetricarboxylic acid chloride, 98%
1,3,5-Benzenetricarboxylic acid chloride, 98%
1,3,5-Benzenetricarboxylic acid chloride, 98%
1,3,5-Benzenetricarboxylic acid chloride, 98%
Thermo Scientific Chemicals
1,3,5-Benzenetricarboxylic acid chloride, 98%
CAS: 4422-95-1 | C9H3Cl3O3 | 265.47 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO164375000500 gGlass bottle
Catalog number ACO164375000
Price (SGD)
1,259.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
1,259.00
EA
Add to cart
Specifications
Chemical Name or Material1, 3, 5-Benzenetricarboxylic acid chloride
CAS4422-95-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0