1x1 image pixel for data collection
Isovaleric acid, 99%
Isovaleric acid, 99%
Isovaleric acid, 99%
Isovaleric acid, 99%
Thermo Scientific Chemicals
Isovaleric acid, 99%
CAS: 503-74-2 | C5H10O2 | 102.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO15669010010 gGlass bottle
Catalog number ACO156690100
Price (SGD)
84.00
EA
Estimated availability date 24-Sep-2025
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
84.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsovaleric acid
Name Note99%
CAS503-74-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
View more
RUO – Research Use Only
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
A0458625Certificate of AnalysisDec 19, 2023156690100, 156691000, FSA156690100, FSA156695000, ACR156690100, ACR156695000, FSA156691000, ACR156691000, 156695000, ACO156690100
A0439314Certificate of AnalysisAug 25, 2023156690100, 156691000, FSA156690100, FSA156695000, ACR156690100, ACR156695000, FSA156691000, ACR156691000, 156695000, ACO156690100
A0454417Certificate of AnalysisAug 25, 2023156690100, 156691000, FSA156690100, FSA156695000, ACR156690100, ACR156695000, FSA156691000, ACR156691000, 156695000, ACO156690100
A0452606Certificate of AnalysisApr 27, 2023156690100, 156691000, FSA156690100, FSA156695000, ACR156690100, ACR156695000, FSA156691000, ACR156691000, 156695000, ACO156690100, 156690025
A0394972Certificate of AnalysisDec 15, 2022156690100, 156691000, FSA156690100, FSA156695000, ACR156690100, ACR156695000, FSA156691000, ACR156691000, 156695000, ACO156690100
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link