Cyclohexene, 99%, pure, stabilized
Cyclohexene, 99%, pure, stabilized
Cyclohexene, 99%, pure, stabilized
Cyclohexene, 99%, pure, stabilized
Thermo Scientific Chemicals
Cyclohexene, 99%, pure, stabilized
CAS: 110-83-8 | C6H10 | 82.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO154845000500 mLGlass bottle
Catalog number ACO154845000
Price (SGD)
65.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
65.00
EA
Add to cart
Specifications
Chemical Name or MaterialCyclohexene
CAS110-83-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi