1-Octanol, 99%, pure
1-Octanol, 99%, pure
1-Octanol, 99%, pure
1-Octanol, 99%, pure
Thermo Scientific Chemicals
1-Octanol, 99%, pure
CAS: 111-87-5 | C8H18O | 130.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO150632500250 mLGlass bottle
Catalog number ACO150632500
Price (SGD)
54.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
54.00
EA
Add to cart
Specifications
Chemical Name or Material1-Octanol
CAS111-87-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0