Ethanolamine, 99%
Ethanolamine, 99%
Ethanolamine, 99%
Thermo Scientific Chemicals
Ethanolamine, 99%
CAS: 141-43-5 | C2H7NO | 61.08 g/mol
Have Questions?
Catalog NumberQuantity
ACO1495800101 L
Catalog number ACO149580010
Price (SGD)
138.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (SGD)
138.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthanolamine
Name Note99%
CAS141-43-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement