Piperidine, 99%, extra pure
Thermo Scientific Chemicals
Piperidine, 99%, extra pure
CAS: 110-89-4 | C5H11N | 85.15 g/mol
Have Questions?
Catalog NumberQuantity
ACO1471800101 L
Catalog number ACO147180010
Price (SGD)
237.00
Each
Quantity:
1 L
Request bulk or custom format
Price (SGD)
237.00
Each
Specifications
Chemical Name or MaterialPiperidine
CAS110-89-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Harmful if swallowed.
Health Hazard 3GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Ground and bond container and receiving equipment Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.
No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.
No smoking.
Ground and bond container and receiving equipment