Sulfuric acid, 96%, extra pure, solution in water
Sulfuric acid, 96%, extra pure, solution in water
Sulfuric acid, 96%, extra pure, solution in water
Thermo Scientific Chemicals
Sulfuric acid, 96%, extra pure, solution in water
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1336100101 LGlass Bottle
Catalog number ACO133610010
Price (SGD)
84.00
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
84.00
Each
Specifications
Chemical Name or MaterialSulfuric acid
ColorColorless
Name Note96% Solution in Water
CAS7664-93-9
Melting Point10.0°C