Propargyl bromide, 80 wt.% solution in toluene, stabilized
Propargyl bromide, 80 wt.% solution in toluene, stabilized
Propargyl bromide, 80 wt.% solution in toluene, stabilized
Propargyl bromide, 80 wt.% solution in toluene, stabilized
Thermo Scientific Chemicals
Propargyl bromide, 80 wt.% solution in toluene, stabilized
CAS: 106-96-7 | C3H3Br | 118.961 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO13148050050 mLGlass bottle
Catalog number ACO131480500
Price (SGD)
137.00
EA
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
137.00
EA
Add to cart
Chemical Identifiers
CAS106-96-7, 108-88-3
Specifications
Chemical Name or MaterialPropargyl bromide
ColorWhite
CAS106-96-7
Boiling Point88.0°C to 90.0°C
Health Hazard 1Danger
View more
Alkylation reagent
RUO – Research Use Only