Piperazine hexahydrate, 98%
Piperazine hexahydrate, 98%
Piperazine hexahydrate, 98%
Piperazine hexahydrate, 98%
Thermo Scientific Chemicals
Piperazine hexahydrate, 98%
CAS: 142-63-2 | C4H10N2·6H2O | 194.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO131305000500 gPlastic bottle
Catalog number ACO131305000
Price (SGD)
150.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
150.00
EA
Add to cart
Specifications
Chemical Name or MaterialPiperazine hexahydrate
Name Note98%
CAS142-63-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility.
Suspected of damaging