m-Phenylenediamine, 99+%
m-Phenylenediamine, 99+%
m-Phenylenediamine, 99+%
m-Phenylenediamine, 99+%
Thermo Scientific Chemicals
m-Phenylenediamine, 99+%
CAS: 108-45-2 | C6H8N2 | 108.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO130565000500 gGlass bottle
Catalog number ACO130565000
Price (SGD)
114.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
114.00
EA
Add to cart
Specifications
Chemical Name or Materialm-Phenylenediamine
CAS108-45-2
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0