n-Pentadecanoic acid, 99%
n-Pentadecanoic acid, 99%
n-Pentadecanoic acid, 99%
Thermo Scientific Chemicals
n-Pentadecanoic acid, 99%
CAS: 1002-84-2 | C15H30O2 | 242.4 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1298300505 gGlass bottle
Catalog number ACO129830050
Price (SGD)
92.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
92.00
Each
Specifications
Chemical Name or Materialn-Pentadecanoic acid
Name Note99%
CAS1002-84-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning