Maleic Acid, 99%
Maleic Acid, 99%
Maleic Acid, 99%
Thermo Scientific Chemicals
Maleic Acid, 99%
CAS: 110-16-7 | C4H4O4 | 116.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO125231000100 gPlastic bottle
Catalog number ACO125231000
Price (SGD)
67.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
67.00
EA
Add to cart
Specifications
Chemical Name or MaterialMaleic acid
CAS110-16-7
Melting Point134.0°C to 138.0°C
ColorWhite
Health Hazard 2GHS H Statement: