Magnesium sulfate heptahydrate, 99+%, extra pure
Magnesium sulfate heptahydrate, 99+%, extra pure
Magnesium sulfate heptahydrate, 99+%, extra pure
Magnesium sulfate heptahydrate, 99+%, extra pure
Thermo Scientific Chemicals
Magnesium sulfate heptahydrate, 99+%, extra pure
CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1249000101 kgPlastic Bottle
Catalog number ACO124900010
Price (SGD)
116.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (SGD)
116.00
EA
Add to cart
Specifications
Chemical Name or MaterialMagnesium sulfate heptahydrate
Melting Point770.0°C
CAS10034-99-8
ColorWhite
pH5.0 to 9.2 (5% soln. at 20°C)