Sodium thiosulfate, 0.1 N standard solution
Sodium thiosulfate, 0.1 N standard solution
Sodium thiosulfate, 0.1 N standard solution
Sodium thiosulfate, 0.1 N standard solution
Thermo Scientific Chemicals
Sodium thiosulfate, 0.1 N standard solution
CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1242700101 LPlastic bottle
Catalog number ACO124270010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7772-98-7, 7732-18-5
Specifications
Chemical Name or MaterialSodium thiosulfate
CAS7772-98-7
ColorColorless
Health Hazard 1
CAS7732-18-5
View more
Reduction reagent, Volumetric standard solution
RUO – Research Use Only