Hydrochloric acid, 1N standard solution
Hydrochloric acid, 1N standard solution
Hydrochloric acid, 1N standard solution
Hydrochloric acid, 1N standard solution
Thermo Scientific Chemicals
Hydrochloric acid, 1N standard solution
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1242100101 LPlastic Bottle
Catalog number ACO124210010
Price (SGD)
76.00
EA
Add to cart
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (SGD)
76.00
EA
Add to cart
Chemical Identifiers
CAS7647-01-0, 7732-18-5
Specifications
Chemical Name or MaterialHydrochloric acid
ColorColorless
Name Note1N standard solution
CAS7647-01-0
Concentration0.998 to 1.002N (20°C)
View more
Volumetric standard solution
RUO – Research Use Only