2-Imidazolidinethione, 98%
2-Imidazolidinethione, 98%
2-Imidazolidinethione, 98%
2-Imidazolidinethione, 98%
Thermo Scientific Chemicals
2-Imidazolidinethione, 98%
CAS: 96-45-7 | C3H6N2S | 102.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1220300101 kgPlastic bottle
Catalog number ACO122030010
Price (SGD)
374.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
374.00
EA
Add to cart
Specifications
Chemical Name or Material2-Imidazolidinethione
Name Note98%
CAS96-45-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger