N,N-Dimethylacetamide, 99%, pure
N,N-Dimethylacetamide, 99%, pure
N,N-Dimethylacetamide, 99%, pure
N,N-Dimethylacetamide, 99%, pure
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1156900101 LGlass Bottle
Catalog number ACO115690010
Price (SGD)
107.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
107.00
EA
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylacetamide
CAS127-19-5
CAS Min %98.5
Health Hazard 1Signal Word: WARNING!
CAS Max %100.0