Diethyl carbonate, 99%
Diethyl carbonate, 99%
Diethyl carbonate, 99%
Diethyl carbonate, 99%
Diethyl carbonate, 99%
Thermo Scientific Chemicals
Diethyl carbonate, 99%
CAS: 105-58-8 | C5H10O3 | 118.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO114221000100 mLGlass bottle
Catalog number ACO114221000
Price (SGD)
48.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
48.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiethyl carbonate
CAS105-58-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
Suitable for battery materials development.
Alkylation reagent
RUO – Research Use Only