Cyclopentane, 97%, extra pure
Cyclopentane, 97%, extra pure
Cyclopentane, 97%, extra pure
Cyclopentane, 97%, extra pure
Thermo Scientific Chemicals
Cyclopentane, 97%, extra pure
Cyclopentane, 97%, C5H10, CAS Number-287-92-3, ciclopentano, cyclopentan, cyclopentanes, cyclopentyl group, hsdb 62, hydrocarbons, cyclic c5 and c6, pentamethylene, t86pb90rnu, unii-t86pb90rnu, zyklopentan, 1L, 96% min. (GC), 05, 19, 50.0 deg.C, CHEBI:23492, White, 0.7500g/mL, 206-016-6, -7 deg.C | CAS: 287-92-3 | C5H10 | 70.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1114800252.5 LGlass bottle
Catalog number ACO111480025
Price (SGD)
763.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
763.00
EA
Add to cart
Specifications
Chemical Name or MaterialCyclopentane
CAS287-92-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o