n-Butyl acetate, 99+%, extra pure
n-Butyl acetate, 99+%, extra pure
n-Butyl acetate, 99+%, extra pure
n-Butyl acetate, 99+%, extra pure
Thermo Scientific Chemicals
n-Butyl acetate, 99+%, extra pure
CAS: 123-86-4 | C6H12O2 | 116.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1077500101 LGlass Bottle
Catalog number ACO107750010
Price (SGD)
118.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
118.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Butyl acetate
CAS123-86-4
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0