L(+)-Ascorbic Acid, 99%
L(+)-Ascorbic Acid, 99%
L(+)-Ascorbic Acid, 99%
L(+)-Ascorbic Acid, 99%
Thermo Scientific Chemicals
L(+)-Ascorbic Acid, 99%
CAS: 50-81-7 | C6H8O6 | 176.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO105021000100 gPlastic bottle
Catalog number ACO105021000
Price (SGD)
72.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
72.00
EA
Add to cart
Specifications
Chemical Name or MaterialL(+)-Ascorbic acid
Name Note99%
CAS50-81-7
Health Hazard 1LIGHT SENSITIVE
Health Hazard 2AIR SENSITIVE