2-Amino-2-methyl-1-propanol, 99%
2-Amino-2-methyl-1-propanol, 99%
2-Amino-2-methyl-1-propanol, 99%
2-Amino-2-methyl-1-propanol, 99%
Thermo Scientific Chemicals
2-Amino-2-methyl-1-propanol, 99%
CAS: 124-68-5 | C4H11NO | 89.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO104065000500 mLGlass bottle
Catalog number ACO104065000
Price (SGD)
106.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
106.00
EA
Add to cart
Specifications
Chemical Name or Material2-Amino-2-methyl-1-propanol
CAS124-68-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0