1,5-Pentanediol, 98%
1,5-Pentanediol, 98%
1,5-Pentanediol, 98%
1,5-Pentanediol, 98%
1,5-Pentanediol, 98%
CAS: 111-29-5 | C5H12O2 | 104.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACG1299500101 kgGlass bottle
Catalog number ACG129950010
Price (SGD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 5-Pentanediol
CAS111-29-5
Melting Point-18.0°C
ColorColorless
Density0.9940g/mL