Heat Seals for ALPS™ Microplate Heat Sealing Instruments
Heat Seals for ALPS™ Microplate Heat Sealing Instruments
Actual product may vary
Heat Seals for ALPS™ Microplate Heat Sealing Instruments
Save time and costs while improving sealing integrity with these microplate heat sealing instruments. Offers the capability of retrieving samples and then re-sealing the plate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialProduct TypeDimensions (L x W)
AB-3797PolyesterClear Seal Thin78 mm x 610 m
AB-3738AluminumEasy Pierce78 mm x 610 m
AB-3739AluminumEasy-Peel78 mm x 610 m
Catalog number AB-3797
Price (SGD)
3,214.00
EA
Add to cart
Material:
Polyester
Product Type:
Clear Seal Thin
Dimensions (L x W):
78 mm x 610 m
Price (SGD)
3,214.00
EA
Add to cart
Save time and costs while improving sealing integrity with Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments. Ideal for storage applications, heat sealing offers the capability of retrieving samples and then re-sealing the plate. When heat sealing, the sealing material is first placed in position over the plate, which must have a raised rim around each well. Heat is applied evenly for several seconds, resulting in binding of the sealing material to the raised rim to achieve a complete seal.
Features:
  • Quick operation
  • Safe for temperature-sensitive samples
  • Designed for reliable performance with Thermo Scientific ALPS instruments, including the, ALPS 3000, ALPS 25 and ALPS 50 V
Specifications
ColorClear
MaterialPolyester
No. of Wells96
No. per Case1
No. per Pack1
Dimensions (L x W)78 mm x 610 m
For Use With (Application)Ideal for fluorescence, colorimetric, and optical applications; suitable for PCR/ qPCR; ELISA
FormatRoll
For Use With (Equipment)ALPS 5000 & ALPS 3000
Product TypeClear Seal Thin
Product LineAbgene
AdhesivePolypropylene
PierceableYes
Temperature Range (Metric)-80°C to +120°C
Plate CompatibilityPolypropylene, Polyethylene, Polystyrene
Incubation ConditionsGeneral
PeelableYes
FluorescenceLow Autofluorescence
Unit SizeEA
Contents & Storage
General; Long term