IP3 Receptor Type II Rabbit anti-Human, Hamster, Mouse, Monkey, Rat, Polyclonal, MilliporeSigma™
IP3 Receptor Type II Rabbit anti-Human, Hamster, Mouse, Monkey, Rat, Polyclonal, MilliporeSigma™
Actual product may vary
IP3 Receptor Type II Rabbit anti-Human, Hamster, Mouse, Monkey, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Have Questions?
Catalog number AB-3000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Anti-IP3 Receptor Type II Antibody detects level of IP3 Receptor Type II and has been published and validated for use in immunocytochemistry, immunofluoresence, immunohistochemistry, immunoprecipitation and western blotting.
For Research Use Only
Specifications
AntigenIP3 Receptor Type II
ApplicationsImmunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
ClassificationPolyclonal
ConjugateUnconjugated
FormulationAffinity purified versus immobilized immunogen peptide. Liquid in PBS buffer, containing 0.05% BSA, 10% glycerol, 0.005% sodium azide. Caution: Sodium azide is hazardous. MSDS available upon request.
Gene SymbolsITPR2; InsP3R2; IP3R2
Host SpeciesRabbit
ImmunogenSynthetic peptide corresponding to the carboxy terminus of the rat Type II receptor
Purification MethodAffinity Purified
Quantity50 μL
Primary or SecondaryPrimary
Gene ID (Entrez)NM_002223.2
Target SpeciesHamster, Human, Monkey, Mouse, Rat
Unit SizeEA
Contents & Storage
−20°C in undiluted aliquotes for 12 months.