Polystyrene 96-Well Plate Cover with Notches
Product Image
Polystyrene 96-Well Plate Cover with Notches
Polystyrene or polypropylene 96-well plate covers, with or without ridges for easy stacking
Have Questions?
Catalog NumberPlate Compatibility
AB-0751Abgene 96 and 384-well Plate
Catalog number AB-0751
Price (SGD)
248.00
CS
Add to cart
Plate Compatibility:
Abgene 96 and 384-well Plate
Price (SGD)
248.00
CS
Add to cart
Polystyrene or polypropylene 96-well plate covers, with or without ridges for easy stacking
Thermo Scientific Polypropylene Plate Covers with stacking ridges provide effective sealing options for a variety of applications and can be used with 96-well plates. The covers are made of polystyrene or polypropylene and are either ridged or non-ridged.
Highlights
  • Polystyrene covers can be used with all 96-well plates
  • Ridged covers for stacking of covered plates
  • Non-ridged covers for use with automated systems
  • Polypropylene covers, ridged and non-ridged, are available for use with and 384-well plates
Hazardous: No
Storage Condition: Ambient
Specifications
ColorClear
DescriptionMicroplate Lid
Dimensions (L x W x H)5 x 3.31 x 0.36 in. (126 x 84 x 9.25mm)
MaterialPolystyrene
SterilityCleanroom
No. of Wells96
IncludesWithout stacking ridge (for automated systems)
Condensation RingsNo
No. per Case50
No. per Pack2
Dimensions (L x W)5 x 3.31 in. (126 x 84 mm)
FormatLid
Product TypeMicroplate Lid, cover
Product LineAbgene
AdhesiveNone
Temperature Range (Metric)-20°C to +120°C
Well ShapeAll well shapes
Plate CompatibilityAbgene 96 and 384-well Plate
Cut-off CornersH1 Corner
Fit for PurposePCR Ready
Unit SizeCS