ALPS Heat Sealer Accessories
Actual product may vary
ALPS Heat Sealer Accessories
ALPS Heat Sealer Accessories
ALPS Heat Sealer Accessories
Thermo Scientific
ALPS Heat Sealer Accessories
Use these ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
AB-0724384-well PCR Plate Carrier
AB-05631000
also known as AB-0563/1000
96-Well PCR Plate Carrier
Catalog number AB-0724
HSN 84229090
Price (SGD)
698.00
Case
Description:
384-well PCR Plate Carrier
Price (SGD)
698.00
Case
Use Thermo Scientific™ ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer. The foil stripper, AB-0592, is also compatible with the ALPS 3000 Microplate Heat Sealer.
 Warranty: 1 year
Specifications
Description384-well PCR Plate Carrier
For Use With (Equipment)ALPS™ Heat Sealer
Product TypeCarrier
Unit SizeCase