2-Part Luer Slip Syringe
2-Part Luer Slip Syringe
Actual product may vary
2-Part Luer Slip Syringe
Inert syringes for use in gas or liquid chromatography, in vitro fertilization, embryo-transfer and various laboratory procedures
Have Questions?
Catalog NumberVolume (Metric)GraduationsConnection Type
A55 mL0.2 ccCentric Luer Slip
Catalog number A5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
5 mL
Graduations:
0.2 cc
Connection Type:
Centric Luer Slip
  • Needle tips not included, purchased separately
  • Graduated syringes manufactured from high-quality laboratory-grade polypropylene and polyethylene
  • Chemically inert and noncytotoxic
  • Ideal for chromatography, in vitro fertilization and embryo transfer
  • Positive safety stop prevents accidental spills
  • FDA registered
Notes:
  • 5 mL syringes graduated to 6 mL
  • 10 mL syringes graduated to 12 mL
  • 20 mL syringes graduated to 24 mL
Over the coming months, you may notice a difference in the box you receive as a branding transition occurs with the Norm-Ject line of syringes. These syringes continue to be manufactured in the same location and the same high standards. Please contact us for more details.
Specifications
Certifications/ComplianceFDA, CE, ISO 9001, DEHP free
MaterialPlastic
For Use With (Application)For nuclear medicine, amniocentesis, IVF, embryo-transfer, chromatography and lab procedures
Latex FreeYes
Product TypeB Braun syringe
Graduations0.2 cc
Quantity100 Pk., 20 Pk./Cs.
PackagingIndividually wrapped
Volume (Metric)5 mL
Product LineB Braun
Needle Point StyleCone
Connection TypeCentric Luer Slip